Assay Details
| Gene Symbol: HD2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120455.4 | NP_195994.3 | 5 - 6 | 616 | 124 | |
| GenBank mRNA | AF255712.1 | - | 4 - 5 | 481 | 124 | |
| AF372889.1 | - | 5 - 6 | 593 | 124 | ||
| AF510168.1 | - | 4 - 5 | 481 | 124 | ||
| AY142039.1 | - | 4 - 5 | 481 | 124 | ||
More Information
Set Membership: |
|

HD2C
Genomic Map