Assay Details
| Gene Symbol: ABCB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120473.3 | NP_196011.1 | 2 - 3 | 737 | 79 | |
| GenBank mRNA | AY059727.1 | - | 2 - 3 | 732 | 79 | |
| AY091365.1 | - | 2 - 3 | 670 | 79 | ||
More Information
Set Membership: |
|

ABCB29
Genomic Map