Assay Details
| Gene Symbol: SLP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335625.1 | NP_001324171.1 | 3 - 4 | 499 | 117 | |
| NM_127474.4 | NP_565447.1 | 3 - 4 | 621 | 117 | ||
| GenBank mRNA | AY051009.1 | - | 3 - 4 | 647 | 117 | |
| AY133826.1 | - | 2 - 3 | 312 | 117 | ||
More Information
Set Membership: |
|

SLP3
Genomic Map