Assay Details
| Gene Symbol: SLP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335625.1 | NP_001324171.1 | 7 - 8 | 1350 | 86 | |
| NM_127474.4 | NP_565447.1 | 7 - 8 | 1472 | 86 | ||
| GenBank mRNA | AY051009.1 | - | 7 - 8 | 1498 | 86 | |
| AY133826.1 | - | 6 - 7 | 1163 | 86 | ||
More Information
Set Membership: |
|

SLP3
Genomic Map