Assay Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084858.1 | NP_001078327.1 | 2 - 3 | 633 | 82 | |
| NM_116050.4 | NP_191744.1 | 2 - 3 | 758 | 82 | ||
| NM_180403.2 | NP_850734.1 | 1 - 2 | 1188 | 82 | ||
| GenBank mRNA | AK117352.1 | - | 1 - 2 | 1188 | 82 | |
| BT033127.1 | - | 2 - 3 | 531 | 82 | ||
| X97941.2 | - | 645 | 82 | |||
More Information
Set Membership: |
|

DAG1
Genomic Map