Assay Details
| Gene Symbol: GG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340213.1 | NP_001325615.1 | 2 - 3 | 298 | 72 | |
| NM_116207.3 | NP_567147.1 | 2 - 3 | 275 | 72 | ||
| NM_180410.1 | NP_850741.1 | 2 - 3 | 230 | 72 | ||
| GenBank mRNA | AF283673.1 | - | 2 - 3 | 230 | 72 | |
| AK175815.1 | - | 2 - 3 | 235 | 72 | ||
| BT024540.1 | - | 2 - 3 | 164 | 72 | ||
More Information
Set Membership: |
|

GG1
Genomic Map