Assay Details
| Gene Symbol: UGP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343490.1 | NP_001330919.1 | 14 - 15 | 1259 | 59 | |
| NM_001343491.1 | NP_001330920.1 | 14 - 15 | 1230 | 59 | ||
| NM_001343492.1 | NP_001330921.1 | 14 - 15 | 1194 | 59 | ||
| NM_001343493.1 | NP_001330922.1 | 14 - 15 | 1113 | 59 | ||
| NM_121737.4 | NP_197233.1 | 17 - 18 | 1358 | 59 | ||
| NM_180506.2 | NP_850837.1 | 13 - 14 | 1226 | 59 | ||
| GenBank mRNA | AF360244.1 | - | 17 - 18 | 1287 | 59 | |
| AF361816.1 | - | 17 - 18 | 1288 | 59 | ||
| AY040042.1 | - | 17 - 18 | 1210 | 59 | ||
More Information
Set Membership: |
|

UGP2
Genomic Map