Assay Details
| Gene Symbol: MYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343542.1 | NP_001331774.1 | 2 - 3 | 122 | 153 | |
| NM_121829.2 | NP_197325.1 | 4 - 5 | 497 | 153 | ||
| NM_180511.1 | NP_850842.1 | 4 - 5 | 497 | 153 | ||
| NM_203068.2 | NP_974797.1 | 4 - 5 | 492 | 153 | ||
| NM_203069.2 | NP_974798.1 | 4 - 5 | 492 | 153 | ||
| NM_203070.2 | NP_974799.1 | 4 - 5 | 492 | 153 | ||
| GenBank mRNA | AF291817.1 | - | 4 - 5 | 427 | 153 | |
More Information
Set Membership: |
|

MYR1
Genomic Map