Assay Details
| Gene Symbol: AGL99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120546.2 | NP_196084.1 | 1 | 984 | 85 | |
| GenBank mRNA | DQ056669.1 | - | 1 | 918 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL99| Gene Symbol: AGL99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120546.2 | NP_196084.1 | 1 | 984 | 85 | |
| GenBank mRNA | DQ056669.1 | - | 1 | 918 | 85 | |
Set Membership: |
|