Assay Details
| Gene Symbol: ADR1-L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120554.2 | NP_196092.1 | 3 - 4 | 1555 | 133 | |
| GenBank mRNA | AY050794.1 | - | 3 - 4 | 1432 | 133 | |
| AY113952.1 | - | 3 - 4 | 1396 | 133 | ||
More Information
Set Membership: |
|

ADR1-L2
Genomic Map