Assay Details
| Gene Symbol: SYP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120658.5 | NP_196195.1 | 4 - 5 | 845 | 62 | |
| GenBank mRNA | AY054184.1 | - | 4 - 5 | 776 | 62 | |
| AY066041.1 | - | 4 - 5 | 658 | 62 | ||
More Information
Set Membership: |
|

SYP31
Genomic Map