Assay Details
| Gene Symbol: BBX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127704.3 | NP_565507.1 | 3 - 4 | 547 | 67 | |
| GenBank mRNA | AY060570.1 | - | 3 - 4 | 477 | 67 | |
| AY125573.1 | - | 3 - 4 | 308 | 67 | ||
More Information
Set Membership: |
|

BBX18
Genomic Map