Assay Details
| Gene Symbol: WSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202976.1 | NP_001189905.1 | 16 - 17 | 2100 | 75 | |
| NM_001338190.1 | NP_001325883.1 | 15 - 16 | 2362 | 75 | ||
| NM_112458.5 | NP_188209.3 | 16 - 17 | 2126 | 75 | ||
| NM_180672.4 | NP_851003.2 | 16 - 17 | 2130 | 75 | ||
More Information
Set Membership: |
|

WSIP2
Genomic Map