Assay Details
| Gene Symbol: WSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202976.1 | NP_001189905.1 | 24 - 25 | 3209 | 85 | |
| NM_001338190.1 | NP_001325883.1 | 23 - 24 | 3471 | 85 | ||
| NM_112458.5 | NP_188209.3 | 24 - 25 | 3235 | 85 | ||
| NM_180672.4 | NP_851003.2 | 24 - 25 | 3239 | 85 | ||
More Information
Set Membership: |
|

WSIP2
Genomic Map