Assay Details
| Gene Symbol: WSIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202976.1 | NP_001189905.1 | 4 - 5 | 568 | 112 | |
| NM_001338190.1 | NP_001325883.1 | 3 - 4 | 800 | 112 | ||
| NM_112458.5 | NP_188209.3 | 4 - 5 | 564 | 112 | ||
| NM_180672.4 | NP_851003.2 | 4 - 5 | 568 | 112 | ||
More Information
Set Membership: |
|

WSIP2
Genomic Map