Assay Details
| Gene Symbol: CPN20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343684.1 | NP_001318614.1 | 3 - 4 | 607 | 88 | |
| NM_001343685.1 | NP_001331898.1 | 3 - 4 | 585 | 88 | ||
| NM_122079.4 | NP_197572.1 | 3 - 4 | 437 | 88 | ||
| NM_180714.4 | NP_851045.1 | 3 - 4 | 447 | 88 | ||
| GenBank mRNA | AB007130.1 | - | 3 - 4 | 414 | 88 | |
| AF268068.1 | - | 2 - 3 | 354 | 88 | ||
| AF428339.1 | - | 3 - 4 | 435 | 88 | ||
| AF428366.1 | - | 3 - 4 | 431 | 88 | ||
| AJ010818.1 | - | 3 - 4 | 405 | 88 | ||
| AY034979.1 | - | 3 - 4 | 435 | 88 | ||
| AY062971.1 | - | 2 - 3 | 354 | 88 | ||
More Information
Set Membership: |
|

CPN20
Genomic Map