Assay Details
| Gene Symbol: HD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122171.3 | NP_197657.3 | 4 - 5 | 475 | 76 | |
| NM_180725.3 | NP_851056.1 | 4 - 5 | 546 | 76 | ||
| GenBank mRNA | AF044914.1 | - | 3 - 4 | 372 | 76 | |
| AF195546.1 | - | 4 - 5 | 432 | 76 | ||
| AY059893.1 | - | 4 - 5 | 446 | 76 | ||
| AY079014.1 | - | 4 - 5 | 444 | 76 | ||
| AY122890.1 | - | 3 - 4 | 372 | 76 | ||
More Information
Set Membership: |
|

HD2B
Genomic Map