Assay Details
| Gene Symbol: HD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180725.3 | NP_851056.1 | 7 - 8 | 888 | 61 | |
| GenBank mRNA | AY059893.1 | - | 7 - 8 | 788 | 61 | |
| AY079014.1 | - | 7 - 8 | 786 | 61 | ||
| AY122890.1 | - | 6 - 7 | 714 | 61 | ||
More Information
Set Membership: |
|

HD2B
Genomic Map