Assay Details
| Gene Symbol: APE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036940.1 | NP_001032017.1 | 2 - 3 | 697 | 133 | |
| NM_001085251.1 | NP_001078720.1 | 2 - 3 | 697 | 133 | ||
| NM_123979.2 | NP_568655.2 | 2 - 3 | 719 | 133 | ||
| NM_180807.2 | NP_851138.1 | 2 - 3 | 712 | 133 | ||
| GenBank mRNA | AF097648.1 | - | 2 - 3 | 354 | 133 | |
| AY037211.1 | - | 2 - 3 | 376 | 133 | ||
| AY050811.1 | - | 2 - 3 | 383 | 133 | ||
| AY091414.1 | - | 2 - 3 | 341 | 133 | ||
| BT002683.1 | - | 2 - 3 | 341 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_123979.2 transcripts.
Set Membership: |
|

APE2
Genomic Map