Assay Details
| Gene Symbol: APE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036940.1 | NP_001032017.1 | 4 - 5 | 847 | 75 | |
| NM_001085251.1 | NP_001078720.1 | 4 - 5 | 847 | 75 | ||
| NM_123979.2 | NP_568655.2 | 4 - 5 | 869 | 75 | ||
| NM_180807.2 | NP_851138.1 | 4 - 5 | 862 | 75 | ||
| GenBank mRNA | AF097648.1 | - | 4 - 5 | 504 | 75 | |
| AY037211.1 | - | 4 - 5 | 526 | 75 | ||
| AY050811.1 | - | 4 - 5 | 533 | 75 | ||
| AY091414.1 | - | 4 - 5 | 491 | 75 | ||
| BT002683.1 | - | 4 - 5 | 491 | 75 | ||
More Information
Set Membership: |
|

APE2
Genomic Map