Assay Details
| Gene Symbol: APE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036940.1 | NP_001032017.1 | 7 - 8 | 1170 | 118 | |
| NM_001085251.1 | NP_001078720.1 | 7 - 8 | 1170 | 118 | ||
| NM_123979.2 | NP_568655.2 | 7 - 8 | 1192 | 118 | ||
| NM_180807.2 | NP_851138.1 | 7 - 8 | 1185 | 118 | ||
| GenBank mRNA | AF097648.1 | - | 7 - 8 | 827 | 118 | |
| AY037211.1 | - | 7 - 8 | 849 | 118 | ||
| AY050811.1 | - | 7 - 8 | 856 | 118 | ||
| AY091414.1 | - | 7 - 8 | 814 | 118 | ||
| BT002683.1 | - | 7 - 8 | 814 | 118 | ||
More Information
Set Membership: |
|

APE2
Genomic Map