Assay Details
| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 2 - 3 | 408 | 67 | |
| NM_180860.2 | NP_851191.1 | 1 - 2 | 651 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fh5| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 2 - 3 | 408 | 67 | |
| NM_180860.2 | NP_851191.1 | 1 - 2 | 651 | 67 | ||
Set Membership: |
|