Assay Details
| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 4 - 5 | 1644 | 96 | |
| NM_180860.2 | NP_851191.1 | 3 - 4 | 1887 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fh5| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 4 - 5 | 1644 | 96 | |
| NM_180860.2 | NP_851191.1 | 3 - 4 | 1887 | 96 | ||
Set Membership: |
|