Assay Details
| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 6 - 7 | 2076 | 122 | |
| NM_180860.2 | NP_851191.1 | 5 - 6 | 2319 | 122 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fh5| Gene Symbol: Fh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124846.2 | NP_200276.1 | 6 - 7 | 2076 | 122 | |
| NM_180860.2 | NP_851191.1 | 5 - 6 | 2319 | 122 | ||
Set Membership: |
|