Assay Details
| Gene Symbol: TRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085301.1 | NP_001078770.1 | 2 - 3 | 452 | 117 | |
| NM_001345351.1 | NP_001331960.1 | 1 - 2 | 462 | 117 | ||
| NM_125334.4 | NP_200751.1 | 2 - 3 | 550 | 117 | ||
| NM_180890.1 | NP_851221.1 | 2 - 3 | 258 | 117 | ||
| GenBank mRNA | AY519544.1 | - | 1 - 2 | 83 | 117 | |
More Information
Set Membership: |
|

TRP1
Genomic Map