Assay Details
| Gene Symbol: TRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085301.1 | NP_001078770.1 | 8 - 9 | 1835 | 80 | |
| NM_001085302.1 | NP_001078771.1 | 8 - 9 | 1834 | 80 | ||
| NM_001345351.1 | NP_001331960.1 | 7 - 8 | 1845 | 80 | ||
| NM_125334.4 | NP_200751.1 | 8 - 9 | 1933 | 80 | ||
| NM_180890.1 | NP_851221.1 | 8 - 9 | 1641 | 80 | ||
| GenBank mRNA | AY519544.1 | - | 7 - 8 | 1466 | 80 | |
More Information
Set Membership: |
|

TRP1
Genomic Map