Assay Details
| Gene Symbol: TRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085301.1 | NP_001078770.1 | 9 - 10 | 1900 | 58 | |
| NM_001085302.1 | NP_001078771.1 | 9 - 10 | 1899 | 58 | ||
| NM_001345351.1 | NP_001331960.1 | 8 - 9 | 1910 | 58 | ||
| NM_125334.4 | NP_200751.1 | 9 - 10 | 1998 | 58 | ||
| NM_180890.1 | NP_851221.1 | 9 - 10 | 1706 | 58 | ||
| GenBank mRNA | AY519544.1 | - | 8 - 9 | 1531 | 58 | |
More Information
Set Membership: |
|

TRP1
Genomic Map