Assay Details
| Gene Symbol: PPAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125571.5 | NP_568939.2 | 4 - 5 | 595 | 80 | |
| NM_180911.2 | NP_851242.1 | 4 - 5 | 528 | 80 | ||
| NM_203247.1 | NP_974976.1 | 4 - 5 | 494 | 80 | ||
| GenBank mRNA | AF361824.1 | - | 4 - 5 | 521 | 80 | |
| AY056064.1 | - | 3 - 4 | 432 | 80 | ||
| AY069910.1 | - | 4 - 5 | 521 | 80 | ||
More Information
Set Membership: |
|

PPAN
Genomic Map