Assay Details
| Gene Symbol: WNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203127.1 | NP_001190056.1 | 5 - 6 | 1137 | 76 | |
| NM_115022.3 | NP_566954.2 | 4 - 5 | 1305 | 76 | ||
| GenBank mRNA | AJ238802.1 | - | 4 - 5 | 1298 | 76 | |
More Information
Set Membership: |
|

WNK5
Genomic Map