Assay Details
| Gene Symbol: ATPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115090.4 | NP_190798.1 | 3 - 4 | 586 | 101 | |
| GenBank mRNA | AY050330.1 | - | 3 - 4 | 396 | 101 | |
| AY086788.1 | - | 3 - 4 | 445 | 101 | ||
| AY094036.1 | - | 3 - 4 | 321 | 101 | ||
More Information
Set Membership: |
|

ATPQ
Genomic Map