Assay Details
| Gene Symbol: RIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339612.1 | NP_001326319.1 | 1 - 2 | 709 | 58 | |
| NM_001339613.1 | NP_001326320.1 | 2 - 3 | 618 | 58 | ||
| NM_001339614.1 | NP_001326317.1 | 2 - 3 | 561 | 58 | ||
| NM_001339615.1 | NP_001326318.1 | 3 - 4 | 514 | 58 | ||
| NM_115195.3 | NP_190903.2 | 3 - 4 | 470 | 58 | ||
More Information
Set Membership: |
|

RIP3
Genomic Map