Assay Details
| Gene Symbol: GCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339630.1 | NP_001325627.1 | 3 - 4 | 757 | 98 | |
| NM_001339631.1 | NP_001325626.1 | 3 - 4 | 755 | 98 | ||
| NM_115236.4 | NP_190944.2 | 3 - 4 | 846 | 98 | ||
| GenBank mRNA | BT012227.1 | - | 3 - 4 | 696 | 98 | |
| BT012412.1 | - | 3 - 4 | 598 | 98 | ||
More Information
Set Membership: |
|

GCP4
Genomic Map