Assay Details
| Gene Symbol: Fes1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339634.1 | NP_001326231.1 | 1 - 2 | 705 | 71 | |
| NM_115240.5 | NP_190948.1 | 2 - 3 | 379 | 71 | ||
| GenBank mRNA | AY087709.1 | - | 2 - 3 | 289 | 71 | |
More Information
Set Membership: |
|

Fes1B
Genomic Map