Assay Details
| Gene Symbol: CDT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115328.4 | NP_191031.1 | 1 - 2 | 221 | 91 | |
| GenBank mRNA | AJ421409.1 | - | 1 - 2 | 148 | 91 | |
| BT029185.1 | - | 1 - 2 | 148 | 91 | ||
More Information
Set Membership: |
|

CDT1B
Genomic Map