Assay Details
| Gene Symbol: AT3G55020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339712.1 | NP_001319752.1 | 12 - 13 | 2149 | 88 | |
| NM_001339713.1 | NP_001326346.1 | 12 - 13 | 2149 | 88 | ||
| NM_001339714.1 | NP_001326345.1 | 12 - 13 | 2149 | 88 | ||
More Information
Set Membership: |
|

AT3G55020
Genomic Map