Assay Details
| Gene Symbol: UBC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115396.4 | NP_567020.1 | 4 - 5 | 803 | 78 | |
| GenBank mRNA | AK317143.1 | - | 4 - 5 | 377 | 78 | |
| AY045883.1 | - | 4 - 5 | 461 | 78 | ||
| AY086490.1 | - | 4 - 5 | 377 | 78 | ||
| AY117262.1 | - | 4 - 5 | 352 | 78 | ||
| DQ027028.1 | - | 4 - 5 | 352 | 78 | ||
| X72625.1 | - | 447 | 78 | |||
More Information
Set Membership: |
|

UBC14
Genomic Map