Assay Details
| Gene Symbol: RBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339730.1 | NP_001326972.1 | 1 - 2 | 417 | 87 | |
| NM_115408.4 | NP_567023.1 | 1 - 2 | 417 | 87 | ||
| GenBank mRNA | AK226780.1 | - | 1 - 2 | 361 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001339730.1 transcripts.
Set Membership: |
|

RBL
Genomic Map