Assay Details
| Gene Symbol: AT3G55580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115415.3 | NP_191117.1 | 4 - 5 | 583 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G55580| Gene Symbol: AT3G55580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115415.3 | NP_191117.1 | 4 - 5 | 583 | 110 | |
Set Membership: |
|