Assay Details
| Gene Symbol: PPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115524.3 | NP_191224.1 | 2 - 3 | 782 | 98 | |
| GenBank mRNA | AK317395.1 | - | 2 - 3 | 654 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPD6| Gene Symbol: PPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115524.3 | NP_191224.1 | 2 - 3 | 782 | 98 | |
| GenBank mRNA | AK317395.1 | - | 2 - 3 | 654 | 98 | |
Set Membership: |
|