Assay Details
| Gene Symbol: AGL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339845.1 | NP_001319775.1 | 3 - 4 | 602 | 81 | |
| NM_001339846.1 | NP_001326285.1 | 2 - 3 | 1722 | 81 | ||
| NM_001339847.1 | NP_001326287.1 | 2 - 3 | 326 | 81 | ||
| NM_001339848.1 | NP_001326286.1 | 1 - 2 | 1286 | 81 | ||
| NM_115583.6 | NP_191282.2 | 3 - 4 | 602 | 81 | ||
| GenBank mRNA | AY141209.1 | - | 2 - 3 | 265 | 81 | |
| BT030046.1 | - | 2 - 3 | 265 | 81 | ||
More Information
Set Membership: |
|

AGL16
Genomic Map