Assay Details
| Gene Symbol: AGL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339846.1 | NP_001326285.1 | 4 - 5 | 1879 | 86 | |
| NM_001339847.1 | NP_001326287.1 | 4 - 5 | 483 | 86 | ||
| NM_001339848.1 | NP_001326286.1 | 3 - 4 | 1443 | 86 | ||
| NM_115583.6 | NP_191282.2 | 5 - 6 | 759 | 86 | ||
| GenBank mRNA | BT030046.1 | - | 4 - 5 | 422 | 86 | |
More Information
Set Membership: |
|

AGL16
Genomic Map