Assay Details
| Gene Symbol: EIF3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115589.5 | NP_567047.1 | 1 - 2 | 460 | 68 | |
| GenBank mRNA | AF285832.1 | - | 1 - 2 | 277 | 68 | |
| AY039913.1 | - | 1 - 2 | 279 | 68 | ||
| AY079357.1 | - | 1 - 2 | 214 | 68 | ||
More Information
Set Membership: |
|

EIF3E
Genomic Map