Assay Details
| Gene Symbol: AGL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339857.1 | NP_001326798.1 | 2 - 3 | 673 | 138 | |
| NM_115599.4 | NP_191298.1 | 2 - 3 | 730 | 138 | ||
| NM_202721.1 | NP_974450.1 | 1 - 2 | 1006 | 138 | ||
| GenBank mRNA | AF312663.1 | - | 2 - 3 | 292 | 138 | |
| AK222220.1 | - | 2 - 3 | 408 | 138 | ||
| BT025774.1 | - | 1 - 2 | 109 | 138 | ||
More Information
Set Membership: |
|

AGL18
Genomic Map