Assay Details
| Gene Symbol: AGL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339857.1 | NP_001326798.1 | 4 - 5 | 830 | 116 | |
| NM_115599.4 | NP_191298.1 | 4 - 5 | 887 | 116 | ||
| NM_202721.1 | NP_974450.1 | 3 - 4 | 1163 | 116 | ||
| GenBank mRNA | AF312663.1 | - | 4 - 5 | 449 | 116 | |
| AK222220.1 | - | 4 - 5 | 565 | 116 | ||
| BT025774.1 | - | 3 - 4 | 266 | 116 | ||
More Information
Set Membership: |
|

AGL18
Genomic Map