Assay Details
| Gene Symbol: AGL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115599.4 | NP_191298.1 | 7 - 8 | 1117 | 69 | |
| NM_202721.1 | NP_974450.1 | 6 - 7 | 1393 | 69 | ||
| GenBank mRNA | AF312663.1 | - | 7 - 8 | 679 | 69 | |
| AK222220.1 | - | 7 - 8 | 795 | 69 | ||
| BT025774.1 | - | 6 - 7 | 496 | 69 | ||
More Information
Set Membership: |
|

AGL18
Genomic Map