Assay Details
| Gene Symbol: AGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035800.1 | NP_001030877.1 | 10 - 11 | 1425 | 99 | |
| NM_001339877.1 | NP_001325466.1 | 10 - 11 | 1239 | 99 | ||
| NM_115615.3 | NP_567051.1 | 10 - 11 | 1536 | 99 | ||
| GenBank mRNA | AF204676.1 | - | 10 - 11 | 1422 | 99 | |
| BT002466.1 | - | 10 - 11 | 1333 | 99 | ||
More Information
Set Membership: |
|

AGK2
Genomic Map