Assay Details
| Gene Symbol: AGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035800.1 | NP_001030877.1 | 5 - 6 | 661 | 84 | |
| NM_001339877.1 | NP_001325466.1 | 5 - 6 | 478 | 84 | ||
| NM_115615.3 | NP_567051.1 | 5 - 6 | 775 | 84 | ||
| GenBank mRNA | AF204676.1 | - | 5 - 6 | 661 | 84 | |
| BT002466.1 | - | 5 - 6 | 572 | 84 | ||
| BT009693.1 | - | 3 - 4 | 328 | 84 | ||
More Information
Set Membership: |
|

AGK2
Genomic Map