Assay Details
| Gene Symbol: THO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123657.3 | NP_568616.1 | 1 - 2 | 651 | 121 | |
| NM_203144.4 | NP_974873.1 | 2 - 3 | 667 | 121 | ||
| GenBank mRNA | AY039925.1 | - | 1 - 2 | 617 | 121 | |
| AY091346.1 | - | 1 - 2 | 188 | 121 | ||
More Information
Set Membership: |
|

THO5
Genomic Map