Assay Details
| Gene Symbol: THO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123657.3 | NP_568616.1 | 4 - 5 | 1578 | 86 | |
| NM_203144.4 | NP_974873.1 | 5 - 6 | 1594 | 86 | ||
| GenBank mRNA | AY039925.1 | - | 4 - 5 | 1544 | 86 | |
| AY091346.1 | - | 4 - 5 | 1115 | 86 | ||
More Information
Set Membership: |
|

THO5
Genomic Map