Assay Details
| Gene Symbol: HAF01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103009.4 | NP_174552.1 | 13 - 14 | 3148 | 115 | |
| GenBank mRNA | AF510669.1 | - | 12 - 13 | 2856 | 115 | |
More Information
Set Membership: |
|

HAF01
Genomic Map